Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGATGTCGGAGTTGCAGAAGGACT[C/T]CATCCAGCTGGACGAGGACAGCGAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610403 | ||||||||||||||||||||
Literature Links: |
CAND2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CAND2 - cullin associated and neddylation dissociated 2 (putative) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001162499.1 | 276 | Missense Mutation | TCC,TTC | S,F 38 | NP_001155971.1 | |
NM_012298.2 | 276 | Missense Mutation | TCC,TTC | S,F 38 | NP_036430.1 | |
XM_011533503.2 | 276 | Missense Mutation | TCC,TTC | S,F 38 | XP_011531805.1 | |
XM_011533504.2 | 276 | Missense Mutation | TCC,TTC | S,F 14 | XP_011531806.1 |