Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTCCATTTCCTCCCTCAGAAGAC[A/G]AAACTGCTAAGGCCAAGGTCCAACA
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 125290 | |||||||||||||||||||||||
Literature Links: |
ALAS1 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
ALAS1 - 5'-aminolevulinate synthase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000688.5 | 535 | Missense Mutation | AAA,GAA | K,E 69 | NP_000679.1 | |
NM_001304443.1 | 535 | Missense Mutation | AAA,GAA | K,E 69 | NP_001291372.1 | |
NM_001304444.1 | 535 | Missense Mutation | AAA,GAA | K,E 86 | NP_001291373.1 | |
NM_199166.2 | 535 | Missense Mutation | AAA,GAA | K,E 69 | NP_954635.1 | |
XM_011533477.1 | 535 | Missense Mutation | AAA,GAA | K,E 86 | XP_011531779.1 | |
XM_011533478.2 | 535 | Missense Mutation | AAA,GAA | K,E 86 | XP_011531780.1 | |
XM_017005873.1 | 535 | Missense Mutation | AAA,GAA | K,E 69 | XP_016861362.1 |