Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTACTTCTTGCTGAGACCCGAGACG[C/G]TTTTCCTGCTGTGCATCAGCTTGGC
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 611931 | |||||||||||||||||||||||
Literature Links: |
PPM1L PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
PPM1L - protein phosphatase, Mg2+/Mn2+ dependent 1L | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317911.1 | 853 | Intron | NP_001304840.1 | |||
NM_001317912.1 | 853 | Intron | NP_001304841.1 | |||
NM_139245.3 | 853 | Missense Mutation | CTT,GTT | L,V 26 | NP_640338.2 | |
XM_011512440.2 | 853 | Missense Mutation | CTT,GTT | L,V 26 | XP_011510742.1 |