Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTTTTTGGATCCCTTTAAATAGCC[G/T]GAACGATCCAGTTTACCAGGTATGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605005 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GALNT7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GALNT7 - polypeptide N-acetylgalactosaminyltransferase 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017423.2 | Intron | NP_059119.2 | ||||
XM_005263062.1 | Intron | XP_005263119.1 | ||||
XM_011532025.2 | Intron | XP_011530327.1 | ||||
XM_017008292.1 | Intron | XP_016863781.1 | ||||
XM_017008293.1 | Intron | XP_016863782.1 |
LOC101930370 - uncharacterized LOC101930370 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |