Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCATCAGCTTCTCCTGACATAAATC[C/T]AGCACATAACATTGTATCAGTCACA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
TMPRSS11B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TMPRSS11B - transmembrane protease, serine 11B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_182502.3 | 1193 | Missense Mutation | AGA,GGA | R,G 353 | NP_872308.2 | |
XM_011531608.2 | 1193 | Missense Mutation | AGA,GGA | R,G 353 | XP_011529910.1 | |
XM_011531609.1 | 1193 | Intron | XP_011529911.1 |