Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCCCTGCTTTACCAAATATTCTTC[A/G]CCAGTACATCTGAAATTAAACAGAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603756 | ||||||||||||||||||||
Literature Links: |
ABCG2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ABCG2 - ATP binding cassette subfamily G member 2 (Junior blood group) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001257386.1 | 2437 | Missense Mutation | GCG,GTG | A,V 607 | NP_001244315.1 | |
NM_004827.2 | 2437 | Silent Mutation | GGC,GGT | G,G 610 | NP_004818.2 | |
XM_005263354.3 | 2437 | Silent Mutation | GGC,GGT | G,G 610 | XP_005263411.1 | |
XM_005263355.3 | 2437 | Silent Mutation | GGC,GGT | G,G 610 | XP_005263412.1 | |
XM_005263356.3 | 2437 | Silent Mutation | GGC,GGT | G,G 608 | XP_005263413.1 | |
XM_011532420.2 | 2437 | Silent Mutation | GGC,GGT | G,G 610 | XP_011530722.1 | |
XM_017008852.1 | 2437 | Silent Mutation | GGC,GGT | G,G 608 | XP_016864341.1 | |
XM_017008853.1 | 2437 | Silent Mutation | GGC,GGT | G,G 610 | XP_016864342.1 |