Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCGCCGGAGGAAGATGAGGCTGAA[A/G]ATTGGGTTCATCTTACGCAGTTTGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605005 | ||||||||||||||||||||
Literature Links: |
GALNT7 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GALNT7 - polypeptide N-acetylgalactosaminyltransferase 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017423.2 | 95 | Silent Mutation | AAA,AAG | K,K 4 | NP_059119.2 | |
XM_005263062.1 | 95 | Silent Mutation | AAA,AAG | K,K 4 | XP_005263119.1 | |
XM_011532025.2 | 95 | Intron | XP_011530327.1 | |||
XM_017008292.1 | 95 | Intron | XP_016863781.1 | |||
XM_017008293.1 | 95 | Intron | XP_016863782.1 |
LOC101930370 - uncharacterized LOC101930370 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |