Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGTCAATGATCTTTCCATCTTTCT[A/T]TGAGGTTCTTGCCTGAGAAGAAGCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605011 | ||||||||||||||||||||
Literature Links: |
ADAMTS3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ADAMTS3 - ADAM metallopeptidase with thrombospondin type 1 motif 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014243.2 | 4124 | Missense Mutation | AAG,TAG | K,* 1186 | NP_055058.2 | |
XM_011532421.2 | 4124 | Nonsense Mutation | AAG,TAG | K,* 1167 | XP_011530723.1 | |
XM_011532422.2 | 4124 | Nonsense Mutation | AAG,TAG | K,* 1158 | XP_011530724.1 |