Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCGGTGGTGAAAAGACCTGCTCCTC[A/G]GACAGCACCAAGACAAGAAAAGCAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602465 | ||||||||||||||||||||
Literature Links: |
SPRY1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SPRY1 - sprouty RTK signaling antagonist 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001258038.1 | 390 | Missense Mutation | CAG,CGG | Q,R 66 | NP_001244967.1 | |
NM_001258039.1 | 390 | Missense Mutation | CAG,CGG | Q,R 66 | NP_001244968.1 | |
NM_005841.2 | 390 | Missense Mutation | CAG,CGG | Q,R 66 | NP_005832.1 | |
NM_199327.2 | 390 | Missense Mutation | CAG,CGG | Q,R 66 | NP_955359.1 | |
XM_005262686.1 | 390 | Missense Mutation | CAG,CGG | Q,R 66 | XP_005262743.1 | |
XM_017007635.1 | 390 | Missense Mutation | CAG,CGG | Q,R 66 | XP_016863124.1 |