Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGACAGATGTATGGCTGACAAGAC[A/G]CTCCCAGAGTGTTCAAAATTACCTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 104145 | ||||||||||||||||||||
Literature Links: |
AFM PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
AFM - afamin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001133.2 | 282 | Silent Mutation | ACA,ACG | T,T 82 | NP_001124.1 | |
XM_017007842.1 | 282 | Silent Mutation | ACA,ACG | T,T 82 | XP_016863331.1 | |
XM_017007843.1 | 282 | Silent Mutation | ACA,ACG | T,T 82 | XP_016863332.1 | |
XM_017007844.1 | 282 | Silent Mutation | ACA,ACG | T,T 82 | XP_016863333.1 |