Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTTCGATTTTGGATGAAATGGCT[A/G]TTACGACAGATGACTGGGAAGTGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610196 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ELMOD2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ELMOD2 - ELMO domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_153702.3 | 552 | Silent Mutation | CTA,CTG | L,L 21 | NP_714913.1 | |
XM_005262885.3 | 552 | Silent Mutation | CTA,CTG | L,L 33 | XP_005262942.1 | |
XM_005262886.2 | 552 | Silent Mutation | CTA,CTG | L,L 21 | XP_005262943.1 | |
XM_011531818.2 | 552 | Silent Mutation | CTA,CTG | L,L 33 | XP_011530120.1 | |
XM_011531819.2 | 552 | Silent Mutation | CTA,CTG | L,L 21 | XP_011530121.1 |