Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCGAGTGCTGGACTCCAATGACAA[C/T]GTGCCCGCTTTCGACCAACCCGTCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608286 | ||||||||||||||||||||
Literature Links: |
LOC101927359 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LOC101927359 - uncharacterized LOC101927359 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PCDH10 - protocadherin 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020815.2 | 1589 | Silent Mutation | AAC,AAT | N,N 246 | NP_065866.1 | |
NM_032961.2 | 1589 | Silent Mutation | AAC,AAT | N,N 246 | NP_116586.1 | |
XM_011532150.1 | 1589 | Silent Mutation | AAC,AAT | N,N 246 | XP_011530452.1 |