Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAAGGTGCTCCCAGGTGACCATGA[A/G]TATCCCAAATGTAGACCCAAGGTAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 614138 | ||||||||||||||||||||
Literature Links: |
RWDD4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RWDD4 - RWD domain containing 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TRAPPC11 - trafficking protein particle complex 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021942.5 | 385 | Silent Mutation | GAA,GAG | E,E 61 | NP_068761.4 | |
NM_199053.2 | 385 | Silent Mutation | GAA,GAG | E,E 61 | NP_951008.1 | |
XM_017008537.1 | 385 | Silent Mutation | GAA,GAG | E,E 61 | XP_016864026.1 | |
XM_017008538.1 | 385 | Silent Mutation | GAA,GAG | E,E 61 | XP_016864027.1 | |
XM_017008539.1 | 385 | UTR 5 | XP_016864028.1 |