Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTTGTGATAACTGCTAGGAATGGA[A/G]TACTGCGGAATGGGTTCCAGCAAGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601194 | ||||||||||||||||||||
Literature Links: |
TLR1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TLR1 - toll like receptor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003263.3 | 3038 | Silent Mutation | TAC,TAT | Y,Y 737 | NP_003254.2 | |
XM_005262662.4 | 3038 | Silent Mutation | TAC,TAT | Y,Y 737 | XP_005262719.1 | |
XM_011513742.2 | 3038 | Silent Mutation | TAC,TAT | Y,Y 737 | XP_011512044.1 | |
XM_011513743.2 | 3038 | Silent Mutation | TAC,TAT | Y,Y 737 | XP_011512045.1 | |
XM_011513744.2 | 3038 | Silent Mutation | TAC,TAT | Y,Y 737 | XP_011512046.1 | |
XM_011513745.2 | 3038 | Silent Mutation | TAC,TAT | Y,Y 737 | XP_011512047.1 | |
XM_017008571.1 | 3038 | Silent Mutation | TAC,TAT | Y,Y 737 | XP_016864060.1 | |
XM_017008572.1 | 3038 | Silent Mutation | TAC,TAT | Y,Y 737 | XP_016864061.1 |