Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAAAAAGAATCTGGCCTTCTTGAG[G/T]TCTAGACTCTATATGCTGGAGAGAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ARHGEF38 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ARHGEF38 - Rho guanine nucleotide exchange factor 38 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001242729.1 | 230 | Missense Mutation | AGG,AGT | R,S 23 | NP_001229658.1 | |
NM_017700.1 | 230 | Missense Mutation | AGG,AGT | R,S 23 | NP_060170.1 | |
XM_006714244.3 | 230 | Intron | XP_006714307.1 | |||
XM_011532050.2 | 230 | Intron | XP_011530352.1 | |||
XM_011532051.2 | 230 | Missense Mutation | AGG,AGT | R,S 23 | XP_011530353.1 |
ARHGEF38-IT1 - ARHGEF38 intronic transcript 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |