Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATAAACAAGTGGAAGGTCATGTTCA[A/G]CTGCAATGGAAACCACAGAGAATCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601275 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GPM6A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GPM6A - glycoprotein M6A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001261447.1 | 1024 | Silent Mutation | NP_001248376.1 | |||
NM_001261448.1 | 1024 | Silent Mutation | NP_001248377.1 | |||
NM_005277.4 | 1024 | Silent Mutation | NP_005268.1 | |||
NM_201591.2 | 1024 | Silent Mutation | NP_963885.1 | |||
NM_201592.2 | 1024 | Silent Mutation | NP_963886.1 | |||
XM_006714183.1 | 1024 | Silent Mutation | XP_006714246.1 | |||
XM_011531877.1 | 1024 | Silent Mutation | XP_011530179.1 | |||
XM_017008036.1 | 1024 | Silent Mutation | XP_016863525.1 |