Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACTTACATTAATTCCATTCAAAAT[A/C]ATCTGTAAATCCAGCAGTAAATGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 147680 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IL2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IL2 - interleukin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000586.3 | 184 | Missense Mutation | ATG,ATT | M,I 43 | NP_000577.2 | |
XM_017008177.1 | 184 | Missense Mutation | ATG,ATT | M,I 43 | XP_016863666.1 |