Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATACGATTTAAAATCGCTGAGGCGC[A/G]TTCAGGAAGAAACCTGTACAAAAAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612127 | ||||||||||||||||||||
Literature Links: |
HSD17B13 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HSD17B13 - hydroxysteroid 17-beta dehydrogenase 13 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136230.2 | 891 | Missense Mutation | CGC,TGC | R,C 240 | NP_001129702.1 | |
NM_178135.4 | 891 | Missense Mutation | CGC,TGC | R,C 276 | NP_835236.2 |
MIR5705 - microRNA 5705 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |