Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCCTTCTCTGTACCCATACAGCAG[C/T]AGCCTAGCAACTCTGCTGGTGATGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 134920 MIM: 606261 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FGF2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FGF2 - fibroblast growth factor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002006.4 | 776 | UTR 3 | NP_001997.5 |
NUDT6 - nudix hydrolase 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007083.4 | 776 | Silent Mutation | CTA,CTG | L,L 270 | NP_009014.2 | |
NM_198041.2 | 776 | Silent Mutation | CTA,CTG | L,L 101 | NP_932158.1 |