Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGACCGGACATTCTGGCTATACCGC[A/G]TCATCATAAAGTTCTTCTGTTTTTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
SDAD1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SDAD1 - SDA1 domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001288983.1 | 2138 | Intron | NP_001275912.1 | |||
NM_001288984.1 | 2138 | Intron | NP_001275913.1 | |||
NM_018115.3 | 2138 | Intron | NP_060585.2 | |||
XM_005263102.4 | 2138 | Missense Mutation | ACG,ATG | T,M 629 | XP_005263159.1 | |
XM_005263105.4 | 2138 | Intron | XP_005263162.1 | |||
XM_017008354.1 | 2138 | Intron | XP_016863843.1 | |||
XM_017008355.1 | 2138 | Intron | XP_016863844.1 |