Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATTCATAGGTAATACTTTGACCCC[A/G]TGTGCATTGATATAGGTGATAAGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM198B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAM198B - family with sequence similarity 198 member B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001031700.2 | 1961 | Silent Mutation | CAC,CAT | H,H 518 | NP_001026870.2 | |
NM_001128424.1 | 1961 | Silent Mutation | CAC,CAT | H,H 510 | NP_001121896.1 | |
NM_016613.6 | 1961 | Silent Mutation | CAC,CAT | H,H 510 | NP_057697.2 |