Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCACCTCTGCGACCTCCTTCTCTGC[C/T]TTCTCTTTGGCCTTCTCCTTCTCCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 134934 MIM: 604407 | ||||||||||||||||||||
Literature Links: |
FGFR3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FGFR3 - fibroblast growth factor receptor 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LETM1 - leucine zipper and EF-hand containing transmembrane protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012318.2 | 2189 | Silent Mutation | AAA,AAG | K,K 729 | NP_036450.1 | |
XM_006713884.1 | 2189 | Silent Mutation | AAA,AAG | K,K 728 | XP_006713947.1 | |
XM_017008187.1 | 2189 | Intron | XP_016863676.1 |