Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTAAACTGCAAGCAGCATCATGGAG[A/T]TTGCAGAATCCTTCCAGGGAAGTCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607406 | ||||||||||||||||||||
Literature Links: |
CCDC158 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CCDC158 - coiled-coil domain containing 158 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
FAM47E-STBD1 - FAM47E-STBD1 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001242939.1 | 1041 | UTR 3 | NP_001229868.1 |
STBD1 - starch binding domain 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003943.4 | 1041 | Missense Mutation | AGA,AGT | R,S 99 | NP_003934.1 |