Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGTGAGCAGCTTCACCAGCAGCAA[C/G]AGCAGCAGCAGGAGGAGACCGCCGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600473 | ||||||||||||||||||||
Literature Links: |
LINC01024 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LINC01024 - long intergenic non-protein coding RNA 1024 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PURA - purine rich element binding protein A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005859.4 | 957 | Missense Mutation | CAG,GAG | Q,E 300 | NP_005850.1 |