Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGCTTCATAGTGGTCCTGCTCCCCG[A/C]ATGCCATTACCTGCTTCTCAGAACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607183 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SEC24A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SEC24A - SEC24 homolog A, COPII coat complex component | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001252231.1 | 258 | Silent Mutation | CGA,CGC | R,R 107 | NP_001239160.1 | |
NM_021982.2 | 258 | Silent Mutation | CGA,CGC | R,R 107 | NP_068817.1 | |
XM_006714523.2 | 258 | Silent Mutation | CGA,CGC | R,R 107 | XP_006714586.1 | |
XM_017008961.1 | 258 | Silent Mutation | CGA,CGC | R,R 81 | XP_016864450.1 | |
XM_017008962.1 | 258 | Intron | XP_016864451.1 | |||
XM_017008963.1 | 258 | Silent Mutation | CGA,CGC | R,R 107 | XP_016864452.1 |