Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATATGGCAGGACTTTGGGATCCCCA[G/T]GGATAATCTCAATGCCTCAGACTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
PLEKHG4B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PLEKHG4B - pleckstrin homology and RhoGEF domain containing G4B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_052909.3 | Intron | NP_443141.3 | ||||
XM_006714446.3 | Intron | XP_006714509.1 | ||||
XM_011513968.2 | Intron | XP_011512270.1 | ||||
XM_011513969.2 | Intron | XP_011512271.1 | ||||
XM_011513970.1 | Intron | XP_011512272.1 | ||||
XM_011513971.2 | Intron | XP_011512273.1 | ||||
XM_011513972.2 | Intron | XP_011512274.1 | ||||
XM_011513973.1 | Intron | XP_011512275.1 | ||||
XM_017009112.1 | Intron | XP_016864601.1 |