Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAGCAGGACTTGGATGTAGTTGCA[A/C]CCCCTGGGCTGGGGTTTCCCCCCGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 180071 MIM: 608886 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PDE6A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PDE6A - phosphodiesterase 6A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000440.2 | 1738 | Missense Mutation | GGT,GTT | G,V 850 | NP_000431.2 | |
XM_011537650.2 | 1738 | Missense Mutation | GGT,GTT | G,V 555 | XP_011535952.1 | |
XM_011537651.2 | 1738 | Missense Mutation | GGT,GTT | G,V 501 | XP_011535953.1 | |
XM_011537652.1 | 1738 | Missense Mutation | GGT,GTT | G,V 491 | XP_011535954.1 | |
XM_011537653.2 | 1738 | Missense Mutation | GGT,GTT | G,V 491 | XP_011535955.1 | |
XM_011537654.2 | 1738 | Missense Mutation | GGT,GTT | G,V 491 | XP_011535956.1 | |
XM_017009572.1 | 1738 | Missense Mutation | GGT,GTT | G,V 769 | XP_016865061.1 |
PPARGC1B - PPARG coactivator 1 beta | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |