Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCCGTAGCCGGCGTCCCCAACATG[A/G]CGGCTCCCCAAGACGTCCACGTCCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603291 | ||||||||||||||||||||
Literature Links: |
BNIP1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
BNIP1 - BCL2 interacting protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001205.2 | 108 | Missense Mutation | ACG,GCG | T,A 2 | NP_001196.2 | |
NM_013978.2 | 108 | Missense Mutation | ACG,GCG | T,A 2 | NP_053581.2 | |
NM_013979.2 | 108 | Missense Mutation | ACG,GCG | T,A 2 | NP_053582.2 | |
NM_013980.2 | 108 | Missense Mutation | ACG,GCG | T,A 2 | NP_053583.2 | |
XM_011534638.1 | 108 | Missense Mutation | ACG,GCG | T,A 2 | XP_011532940.1 | |
XM_011534639.1 | 108 | Missense Mutation | ACG,GCG | T,A 2 | XP_011532941.1 |
CREBRF - CREB3 regulatory factor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |