Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTCATCTTCGGCATGTTCGGCATC[A/G]TGGTCATGGTCATCGAGACCGAGCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605879 | ||||||||||||||||||||
Literature Links: |
KCNN2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KCNN2 - potassium calcium-activated channel subfamily N member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278204.1 | 2179 | UTR 5 | NP_001265133.1 | |||
NM_021614.3 | 2179 | Missense Mutation | ATG,GTG | M,V 147 | NP_067627.2 | |
NM_170775.2 | 2179 | Intron | NP_740721.1 | |||
XM_011543387.1 | 2179 | Missense Mutation | ATG,GTG | M,V 425 | XP_011541689.1 | |
XM_011543389.1 | 2179 | Missense Mutation | ATG,GTG | M,V 425 | XP_011541691.1 | |
XM_017009457.1 | 2179 | Intron | XP_016864946.1 |