Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCATGTTTCATTTTCCTGAAGATTT[A/G]TGTTTTTGTCTACCTTGTGAGCAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600783 MIM: 612695 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HARS2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HARS2 - histidyl-tRNA synthetase 2, mitochondrial | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
VTRNA1-1 - vault RNA 1-1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZMAT2 - zinc finger matrin-type 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_144723.2 | 1445 | UTR 3 | NP_653324.1 |