Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAAATATACACCCTAAAAGGCCCA[C/T]GATGCTAATCATGATGTCCTTGGCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 613109 MIM: 608332 | ||||||||||||||||||||
Literature Links: |
GM2A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GM2A - GM2 ganglioside activator | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC36A3 - solute carrier family 36 member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145017.1 | 1561 | Missense Mutation | ATG,GTG | M,V 479 | NP_001138489.1 | |
NM_181774.3 | 1561 | Missense Mutation | ATG,GTG | M,V 438 | NP_861439.3 | |
XM_011537627.2 | 1561 | Missense Mutation | ATG,GTG | M,V 412 | XP_011535929.1 | |
XM_011537629.2 | 1561 | UTR 3 | XP_011535931.1 | |||
XM_011537630.1 | 1561 | Missense Mutation | ATG,GTG | M,V 323 | XP_011535932.1 | |
XM_011537631.1 | 1561 | Missense Mutation | ATG,GTG | M,V 323 | XP_011535933.1 | |
XM_011537633.1 | 1561 | Missense Mutation | ATG,GTG | M,V 275 | XP_011535935.1 | |
XM_011537634.2 | 1561 | Intron | XP_011535936.1 |