Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTGCTGGTCCCTCTCGGAGGCGTT[A/G]GACAGGGAGCTGAGGTTGGACGACG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
BRD9 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
BRD9 - bromodomain containing 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001009877.2 | 1656 | Silent Mutation | TCC,TCT | S,S 501 | NP_001009877.2 | |
NM_001317951.1 | 1656 | Silent Mutation | TCC,TCT | S,S 458 | NP_001304880.1 | |
NM_023924.4 | 1656 | Silent Mutation | TCC,TCT | S,S 554 | NP_076413.3 | |
XM_006714494.3 | 1656 | Intron | XP_006714557.1 | |||
XM_017009780.1 | 1656 | Intron | XP_016865269.1 |
ZDHHC11 - zinc finger DHHC-type containing 11 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |