Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCAAAAATGGCGGACGCATTCGGA[A/G]ATGAGCTGTTCAGCGTGTTCGAGGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612720 | ||||||||||||||||||||
Literature Links: |
DHX29 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DHX29 - DEAH-box helicase 29 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_019030.2 | 285 | Intron | NP_061903.2 | |||
XM_006714653.2 | 285 | Intron | XP_006714716.1 | |||
XM_017009588.1 | 285 | Intron | XP_016865077.1 |
SKIV2L2 - Ski2 like RNA helicase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015360.4 | 285 | Missense Mutation | AAT,GAT | N,D 7 | NP_056175.3 |