Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCATATGTTAATACTATATGGTGAA[C/T]GCAAGACAGGGTAAATTTTACTGTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604677 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
COL4A3BP PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
COL4A3BP - collagen type IV alpha 3 binding protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130105.1 | Intron | NP_001123577.1 | ||||
NM_005713.2 | Intron | NP_005704.1 | ||||
NM_031361.2 | Intron | NP_112729.1 | ||||
XM_006714513.2 | Intron | XP_006714576.1 | ||||
XM_011543090.2 | Intron | XP_011541392.1 | ||||
XM_011543091.2 | Intron | XP_011541393.1 | ||||
XM_017008919.1 | Intron | XP_016864408.1 |