Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTAAGGGTCGGGCATTCAGTTCTTT[C/G]AGAGATATACTCCCCTAGAATAGAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607690 | ||||||||||||||||||||
Literature Links: |
SAR1B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SAR1B - secretion associated Ras related GTPase 1B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001033503.2 | 629 | Silent Mutation | CTC,CTG | L,L 165 | NP_001028675.1 | |
NM_016103.3 | 629 | Silent Mutation | CTC,CTG | L,L 165 | NP_057187.1 | |
XM_017009521.1 | 629 | Silent Mutation | CTC,CTG | L,L 165 | XP_016865010.1 |