Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAATCTTGTAGATGATCCAGTAGAA[A/C]ATGTTGAAAATGAGGAAGGCCATGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 138491 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GLRA1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GLRA1 - glycine receptor alpha 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000171.3 | 1583 | Missense Mutation | ATG,ATT | M,I 432 | NP_000162.2 | |
NM_001146040.1 | 1583 | Missense Mutation | ATG,ATT | M,I 440 | NP_001139512.1 | |
NM_001292000.1 | 1583 | Missense Mutation | ATG,ATT | M,I 349 | NP_001278929.1 | |
XM_017009349.1 | 1583 | Missense Mutation | ATG,ATT | M,I 448 | XP_016864838.1 |