Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCCCCAAGACGTCCACGTCCGGAT[C/T]TGTAACCAAGAGATTGTCAAATTTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603291 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BNIP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BNIP1 - BCL2 interacting protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001205.2 | 137 | Silent Mutation | ATC,ATT | I,I 11 | NP_001196.2 | |
NM_013978.2 | 137 | Silent Mutation | ATC,ATT | I,I 11 | NP_053581.2 | |
NM_013979.2 | 137 | Silent Mutation | ATC,ATT | I,I 11 | NP_053582.2 | |
NM_013980.2 | 137 | Silent Mutation | ATC,ATT | I,I 11 | NP_053583.2 | |
XM_011534638.1 | 137 | Silent Mutation | ATC,ATT | I,I 11 | XP_011532940.1 | |
XM_011534639.1 | 137 | Silent Mutation | ATC,ATT | I,I 11 | XP_011532941.1 |
CREBRF - CREB3 regulatory factor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |