Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAATATCGTGAAGGATTTCAGAGG[G/T]TTTTTTCAATGCTATTGATACTCTA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
C5orf34 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C5orf34 - chromosome 5 open reading frame 34 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_198566.3 | 2207 | Missense Mutation | ACC,CCC | T,P 622 | NP_940968.1 | |
XM_011514038.2 | 2207 | UTR 3 | XP_011512340.1 | |||
XM_017009445.1 | 2207 | Missense Mutation | ACC,CCC | T,P 508 | XP_016864934.1 | |
XM_017009446.1 | 2207 | Missense Mutation | ACC,CCC | T,P 508 | XP_016864935.1 |
TMEM267 - transmembrane protein 267 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |