Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAAGTATGTTTGCCTAAAGTCACA[C/T]AATCTGTGAGGCACTGAGCCAGGAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608212 MIM: 612429 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IRGM PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IRGM - immunity related GTPase M | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145805.1 | 2930 | Intron | NP_001139277.1 | |||
XM_011537641.2 | 2930 | Intron | XP_011535943.1 |
ZNF300 - zinc finger protein 300 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001172831.1 | 2930 | UTR 3 | NP_001166302.1 | |||
NM_001172832.1 | 2930 | UTR 3 | NP_001166303.1 | |||
NM_052860.2 | 2930 | UTR 3 | NP_443092.1 | |||
XM_011537702.2 | 2930 | UTR 3 | XP_011536004.1 |