Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATATTACCATCACAGCCTGATTACA[A/C]TAGCAGTAAAATCCCTTCCGCTATG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604103 | ||||||||||||||||||||
Literature Links: |
MYOT PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MYOT - myotilin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135940.1 | 632 | Intron | NP_001129412.1 | |||
NM_001300911.1 | 632 | UTR 5 | NP_001287840.1 | |||
NM_006790.2 | 632 | Missense Mutation | AAT,ACT | N,T 108 | NP_006781.1 | |
XM_017010060.1 | 632 | UTR 5 | XP_016865549.1 | |||
XM_017010061.1 | 632 | UTR 5 | XP_016865550.1 | |||
XM_017010062.1 | 632 | Intron | XP_016865551.1 |