Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGCACGCCCTCCTCCATGTAGAAG[A/G]CCCTGTTCCGGATGGGCGTCTTGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 126660 MIM: 605903 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DBN1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DBN1 - drebrin 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PDLIM7 - PDZ and LIM domain 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005451.4 | 1222 | Missense Mutation | GCC,GTC | A,V 378 | NP_005442.2 | |
NM_203352.2 | 1222 | Missense Mutation | GCC,GTC | A,V 344 | NP_976227.1 | |
NM_213636.2 | 1222 | Intron | NP_998801.1 | |||
XM_011534698.2 | 1222 | Missense Mutation | GCC,GTC | A,V 285 | XP_011533000.1 | |
XM_011534699.2 | 1222 | Missense Mutation | GCC,GTC | A,V 230 | XP_011533001.1 |