Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCAGGAACTGCTCTCAAACCTGGG[C/T]CACCCGGAGAGCCCCCCACGGAAAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601436 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LMBRD2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LMBRD2 - LMBR1 domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001007527.1 | 315 | Intron | NP_001007528.1 | |||
XM_011514162.2 | 315 | Intron | XP_011512464.1 | |||
XM_017010024.1 | 315 | Intron | XP_016865513.1 |
MIR580 - microRNA 580 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SKP2 - S-phase kinase-associated protein 2, E3 ubiquitin protein ligase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243120.1 | 315 | UTR 5 | NP_001230049.1 | |||
NM_005983.3 | 315 | Silent Mutation | GGC,GGT | G,G 60 | NP_005974.2 | |
NM_032637.3 | 315 | Silent Mutation | GGC,GGT | G,G 60 | NP_116026.1 | |
XM_011514081.2 | 315 | Silent Mutation | GGC,GGT | G,G 60 | XP_011512383.1 | |
XM_011514082.2 | 315 | Silent Mutation | GGC,GGT | G,G 60 | XP_011512384.1 | |
XM_011514083.2 | 315 | UTR 5 | XP_011512385.1 | |||
XM_017009753.1 | 315 | UTR 5 | XP_016865242.1 |