Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGAGAAAAAAAGTTTAAGTCAACC[A/G]CTGTTACAATAAAAAAATTTCAGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 114080 MIM: 607049 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CAMK4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CAMK4 - calcium/calmodulin dependent protein kinase IV | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
STARD4 - StAR related lipid transfer domain containing 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308056.1 | Intron | NP_001294985.1 | ||||
NM_001308057.1 | Intron | NP_001294986.1 | ||||
NM_001308058.1 | Intron | NP_001294987.1 | ||||
NM_001308059.1 | Intron | NP_001294988.1 | ||||
NM_001308060.1 | Intron | NP_001294989.1 | ||||
NM_001308061.1 | Intron | NP_001294990.1 | ||||
NM_139164.2 | Intron | NP_631903.1 | ||||
XM_017009041.1 | Intron | XP_016864530.1 | ||||
XM_017009042.1 | Intron | XP_016864531.1 | ||||
XM_017009043.1 | Intron | XP_016864532.1 | ||||
XM_017009044.1 | Intron | XP_016864533.1 |