Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCTCTCAGACCCAGATGTGACGGG[A/G]TGTGTGGCCCGAGGAAGCTGGACAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601530 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C5orf45 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C5orf45 - chromosome 5 open reading frame 45 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001017987.2 | 749 | Silent Mutation | CAC,CAT | H,H 235 | NP_001017987.1 | |
NM_016175.3 | 749 | Silent Mutation | CAC,CAT | H,H 290 | NP_057259.2 |
SQSTM1 - sequestosome 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142298.1 | 749 | UTR 3 | NP_001135770.1 | |||
NM_001142299.1 | 749 | UTR 3 | NP_001135771.1 | |||
NM_003900.4 | 749 | UTR 3 | NP_003891.1 | |||
XM_011534683.2 | 749 | Intron | XP_011532985.1 | |||
XM_017010010.1 | 749 | Intron | XP_016865499.1 |