Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATTTAAGAAATCCCAAATCAAAAT[A/G]GTGTCATCATGGGAGCTGCTGATGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605651 | ||||||||||||||||||||
Literature Links: |
FBXW11 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FBXW11 - F-box and WD repeat domain containing 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012300.2 | 1468 | Silent Mutation | ACC,ACT | T,T 513 | NP_036432.2 | |
NM_033644.2 | 1468 | Silent Mutation | ACC,ACT | T,T 500 | NP_387448.2 | |
NM_033645.2 | 1468 | Silent Mutation | ACC,ACT | T,T 479 | NP_387449.2 | |
XM_005265855.4 | 1468 | Silent Mutation | ACC,ACT | T,T 534 | XP_005265912.1 | |
XM_005265856.2 | 1468 | Silent Mutation | ACC,ACT | T,T 502 | XP_005265913.1 | |
XM_005265857.2 | 1468 | Silent Mutation | ACC,ACT | T,T 486 | XP_005265914.1 | |
XM_005265858.2 | 1468 | Silent Mutation | ACC,ACT | T,T 481 | XP_005265915.1 | |
XM_017009279.1 | 1468 | Silent Mutation | ACC,ACT | T,T 513 | XP_016864768.1 | |
XM_017009280.1 | 1468 | Silent Mutation | ACC,ACT | T,T 481 | XP_016864769.1 | |
XM_017009281.1 | 1468 | Silent Mutation | ACC,ACT | T,T 481 | XP_016864770.1 |