Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGCTCGGGTGGTTTCTAATGATAA[A/C]AAAGAGTGTTTGCAGCTGGACACAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606337 MIM: 606338 MIM: 606339 | ||||||||||||||||||||
Literature Links: |
PCDHB11 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PCDHB11 - protocadherin beta 11 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PCDHB12 - protocadherin beta 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018932.3 | 402 | Missense Mutation | AAA,AAC | K,N 71 | NP_061755.1 |
PCDHB13 - protocadherin beta 13 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |