Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGGACACGGCTCCCACCTCCAGGT[A/C]CAGGGCCACCCGCACGCGCGACAGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609315 | ||||||||||||||||||||
Literature Links: |
LOC102577426 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LOC102577426 - uncharacterized LOC102577426 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TRIM7 - tripartite motif containing 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_033342.3 | 1470 | Intron | NP_203128.1 | |||
NM_203293.2 | 1470 | Missense Mutation | NP_976038.1 | |||
NM_203294.1 | 1470 | Missense Mutation | NP_976039.1 | |||
NM_203295.1 | 1470 | Missense Mutation | NP_976040.1 | |||
NM_203296.1 | 1470 | Missense Mutation | NP_976041.1 | |||
NM_203297.1 | 1470 | Missense Mutation | NP_976042.1 | |||
XM_017009903.1 | 1470 | Missense Mutation | XP_016865392.1 | |||
XM_017009904.1 | 1470 | Intron | XP_016865393.1 |