Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAATGCAAATGCTGGATACAGTGTA[C/T]GTGAACAGTCCACTTGGCGTTCATA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609317 | ||||||||||||||||||||
Literature Links: |
TRIM36 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TRIM36 - tripartite motif containing 36 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001017397.1 | 2611 | Intron | NP_001017397.1 | |||
NM_001017398.1 | 2611 | Intron | NP_001017398.1 | |||
NM_001300752.1 | 2611 | Missense Mutation | CAT,CGT | H,R 542 | NP_001287681.1 | |
NM_001300759.1 | 2611 | Missense Mutation | CAT,CGT | H,R 685 | NP_001287688.1 | |
NM_018700.3 | 2611 | Missense Mutation | CAT,CGT | H,R 697 | NP_061170.2 | |
XM_017009621.1 | 2611 | Missense Mutation | CAT,CGT | H,R 714 | XP_016865110.1 | |
XM_017009622.1 | 2611 | Missense Mutation | CAT,CGT | H,R 702 | XP_016865111.1 | |
XM_017009623.1 | 2611 | Intron | XP_016865112.1 |