Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGCAAGTAGTCCAAATTTAGATTT[C/T]ATAAATCACTTTCAATTTATTAAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602547 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR548P PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MIR548P - microRNA 548p | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ST8SIA4 - ST8 alpha-N-acetyl-neuraminide alpha-2,8-sialyltransferase 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005668.5 | 3595 | UTR 3 | NP_005659.1 | |||
NM_175052.2 | 3595 | Intron | NP_778222.1 | |||
XM_005272078.2 | 3595 | Intron | XP_005272135.1 | |||
XM_011543630.1 | 3595 | UTR 3 | XP_011541932.1 |