Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCAAGTAAGTTCATGACTCACGTG[A/G]CCTCCCAGTTTGCCTCCAGCTATGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
LSM11 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LSM11 - LSM11, U7 small nuclear RNA associated | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
THG1L - tRNA-histidine guanylyltransferase 1 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317824.1 | 440 | Intron | NP_001304753.1 | |||
NM_001317825.1 | 440 | Missense Mutation | ACC,GCC | T,A 5 | NP_001304754.1 | |
NM_001317826.1 | 440 | Missense Mutation | ACC,GCC | T,A 68 | NP_001304755.1 | |
NM_017872.4 | 440 | Missense Mutation | ACC,GCC | T,A 130 | NP_060342.2 | |
XM_017009603.1 | 440 | Intron | XP_016865092.1 |